Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022701.2 | NP_073192.2 | 7 - 8 | 740 | 65 | |
XM_006256024.2 | XP_006256086.1 | 7 - 8 | 759 | 65 | ||
GenBank mRNA | BC064652.1 | - | 7 - 8 | 739 | 5598180 | 65 |
DY569467.1 | - | 4 - 5 | 422 | 65 | ||
U60976.1 | - | 7 - 8 | 670 | 65 | ||
U60977.1 | - | 6 - 7 | 744 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |