Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212498.1 | NP_997663.1 | 9 - 10 | 737 | 128 | |
XM_006255948.2 | XP_006256010.1 | 8 - 9 | 703 | 128 | ||
XM_006255950.2 | XP_006256012.1 | 9 - 10 | 772 | 128 | ||
XM_006255951.2 | XP_006256013.1 | 8 - 9 | 703 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |