Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270855.1 | NP_001257784.1 | 3 - 4 | 290 | 81 | |
NM_001270859.1 | NP_001257788.1 | 3 - 4 | 221 | 81 | ||
NM_019176.2 | NP_062049.1 | 3 - 4 | 290 | 81 | ||
XM_006252149.2 | XP_006252211.1 | 4 - 5 | 444 | 81 | ||
XM_006252151.2 | XP_006252213.1 | 3 - 4 | 1353 | 81 | ||
XM_008770778.1 | XP_008769000.1 | 3 - 4 | 297 | 81 | ||
GenBank mRNA | AF026530.1 | - | 3 - 4 | 231 | 81 | |
BC092646.1 | - | 3 - 4 | 227 | 7319130 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |