Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014051.1 | NP_001014073.1 | 13 - 14 | 1213 | 99 | |
XM_006235276.2 | XP_006235338.1 | 13 - 14 | 1984 | 99 | ||
XM_008762331.1 | XP_008760553.1 | 14 - 15 | 1268 | 99 | ||
XM_008762332.1 | XP_008760554.1 | 14 - 15 | 1268 | 99 | ||
XM_008762333.1 | XP_008760555.1 | 13 - 14 | 1082 | 99 | ||
GenBank mRNA | BC082105.1 | - | 13 - 14 | 1213 | 7134735 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |