Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175593.2 | NP_783183.1 | 2 - 3 | 225 | 136 | |
GenBank mRNA | AY196208.1 | - | 2 - 3 | 225 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |