Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198749.2 | NP_942044.1 | 2 - 3 | 564 | 86 | |
GenBank mRNA | BC094954.1 | - | 2 - 3 | 564 | 7316003 | 86 |
M83679.1 | - | 2 - 3 | 408 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |