Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012751.1 | NP_036883.1 | 1 - 2 | 172 | 89 | |
XM_006246596.2 | XP_006246658.1 | 2 - 3 | 145 | 89 | ||
GenBank mRNA | BC085757.1 | - | 1 - 2 | 217 | 7192278 | 89 |
D28561.1 | - | 1 - 2 | 172 | 89 | ||
FQ216450.1 | - | 2 - 3 | 111 | 89 | ||
FQ232657.1 | - | 1 - 2 | 231 | 89 | ||
FQ233419.1 | - | 1 - 2 | 207 | 89 | ||
J04524.1 | - | 1 - 2 | 154 | 89 | ||
KJ696745.1 | - | 1 - 2 | 57 | 89 | ||
M25482.1 | - | 1 - 2 | 137 | 89 | ||
X14771.1 | - | 1 - 2 | 185 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |