Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007732.1 | NP_001007733.1 | 6 - 7 | 894 | 79 | |
XM_006253872.2 | XP_006253934.1 | 6 - 7 | 863 | 79 | ||
GenBank mRNA | BC079063.1 | - | 6 - 7 | 894 | 7121006 | 79 |
FQ226618.1 | - | 3 - 4 | 429 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |