Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257554.2 | XP_006257616.2 | 1 - 2 | 246 | 80 | |
XM_229133.8 | XP_229133.5 | 1 - 2 | 246 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |