Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130583.1 | NP_001124055.1 | 9 - 10 | 778 | 70 | |
XM_006241411.1 | XP_006241473.1 | 9 - 10 | 735 | 70 | ||
XM_006241412.2 | XP_006241474.1 | 9 - 10 | 988 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |