Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nppb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031545.1 | NP_113733.1 | 1 - 2 | 185 | 94 | |
GenBank mRNA | FQ228997.1 | - | 241 | 94 | ||
M25297.1 | - | 1 - 2 | 185 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |