Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lilra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076793.1 | NP_001070261.1 | 2 - 3 | 69 | 106 | |
GenBank mRNA | AY998481.1 | - | 2 - 3 | 69 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |