Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191814.1 | NP_001178743.1 | 7 - 8 | 1272 | 100 | |
XM_006245658.2 | XP_006245720.1 | 8 - 9 | 1930 | 100 | ||
XM_006245661.2 | XP_006245723.1 | 7 - 8 | 1868 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |