Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201270.1 | NP_958427.1 | 2 - 3 | 213 | 85 | |
GenBank mRNA | X16058.2 | - | 2 - 3 | 213 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Rattus norvegicus Inventoried Probe spans exons |