Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001183551.3 | NP_014775.3 | 1 | 1553 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |