Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005660442.3 | XP_005660499.1 | 229 | 94 | ||
GenBank mRNA | AK237253.1 | - | 2 - 3 | 199 | 94 | |
AK345030.1 | - | 2 - 3 | 207 | 94 | ||
AK394515.1 | - | 2 - 3 | 203 | 94 | ||
AK394884.1 | - | 1 - 2 | 766 | 94 | ||
AK397580.1 | - | 2 - 3 | 199 | 94 | ||
AY550045.1 | - | 2 - 3 | 189 | 94 | ||
AY610349.1 | - | 2 - 3 | 204 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |