Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213853.1 | NP_999018.1 | 5 | 649 | 90 | |
XM_005653648.3 | XP_005653705.1 | 900 | 90 | |||
XM_005653650.2 | XP_005653707.1 | 911 | 90 | |||
XM_005653651.3 | XP_005653708.1 | 596 | 90 | |||
XM_005653652.2 | XP_005653709.1 | 658 | 90 | |||
XM_005653653.2 | XP_005653710.1 | 689 | 90 | |||
XM_005653654.3 | XP_005653711.1 | 936 | 90 | |||
XM_021102177.1 | XP_020957836.1 | 833 | 90 | |||
GenBank mRNA | AK236612.1 | - | 5 | 766 | 90 | |
AK397590.1 | - | 5 | 698 | 90 | ||
AK398529.1 | - | 5 | 729 | 90 | ||
U89949.1 | - | 5 | 649 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |