Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097459.1 | NP_001090928.1 | 9 - 10 | 2229 | 98 | |
XM_005673829.3 | XP_005673886.1 | 3217 | 98 | |||
GenBank mRNA | EF101315.1 | - | 9 - 10 | 2229 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |