Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HADHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348972.1 | NP_001335901.1 | 8 - 9 | 770 | 99 | |
GenBank mRNA | AF028007.1 | - | 5 - 6 | 534 | 99 | |
AK239646.1 | - | 8 - 9 | 717 | 99 | ||
AK343679.1 | - | 8 - 9 | 730 | 99 | ||
AK389670.1 | - | 8 - 9 | 738 | 99 | ||
AK393956.1 | - | 8 - 9 | 706 | 99 | ||
AK400872.1 | - | 8 - 9 | 691 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |