Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244893.1 | NP_001231822.1 | 2 - 3 | 741 | 78 | |
GenBank mRNA | AK236177.1 | - | 2 - 3 | 705 | 78 | |
AK237169.1 | - | 2 - 3 | 741 | 78 | ||
AK395565.1 | - | 2 - 3 | 741 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |