Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244980.1 | NP_001231909.1 | 5 - 6 | 534 | 102 | |
XM_005674019.3 | XP_005674076.1 | 637 | 102 | |||
GenBank mRNA | AK230843.1 | - | 5 - 6 | 534 | 102 | |
AK392033.1 | - | 5 - 6 | 547 | 102 | ||
AK399546.1 | - | 5 - 6 | 557 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |