Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206447.1 | NP_001193376.1 | 1 - 2 | 1060 | 96 | |
XM_013981389.2 | XP_013836843.2 | 1325 | 96 | |||
GenBank mRNA | AK351123.1 | - | 1 - 2 | 1058 | 96 | |
AK389431.1 | - | 1 - 2 | 487 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |