Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243412.1 | NP_001230341.1 | 8 - 9 | 1062 | 105 | |
NM_001243413.1 | NP_001230342.1 | 7 - 8 | 919 | 105 | ||
XM_005655700.3 | XP_005655757.1 | 1702 | 105 | |||
XM_013988430.2 | XP_013843884.1 | 1787 | 105 | |||
XM_013988431.2 | XP_013843885.1 | 961 | 105 | |||
XM_021090919.1 | XP_020946578.1 | 1708 | 105 | |||
XM_021090921.1 | XP_020946580.1 | 951 | 105 | |||
XM_021090922.1 | XP_020946581.1 | 1787 | 105 | |||
XM_021090923.1 | XP_020946582.1 | 776 | 105 | |||
XM_021090924.1 | XP_020946583.1 | 1513 | 105 | |||
GenBank mRNA | AK400126.1 | - | 8 - 9 | 1045 | 105 | |
BP443627.1 | - | 4 - 5 | 447 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |