Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243883.1 | NP_001230812.1 | 7 - 8 | 1143 | 101 | |
GenBank mRNA | AJ236934.1 | - | 7 - 8 | 1103 | 101 | |
AK231339.1 | - | 7 - 8 | 1143 | 101 | ||
AK237000.1 | - | 7 - 8 | 1109 | 101 | ||
AK344422.1 | - | 7 - 8 | 942 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |