Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP1;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ834694.1 | - | 2 - 3 | 655 | 86 | |
EF364432.1 | - | 2 - 3 | 642 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Vitis vinifera Probe spans exons |