Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004983.1 | NP_001004983.1 | 6 - 7 | 591 | 68 | |
XM_012970297.2 | XP_012825751.1 | 6 - 7 | 650 | 68 | ||
XM_012970298.2 | XP_012825752.1 | 6 - 7 | 476 | 68 | ||
XM_018097081.1 | XP_017952570.1 | 5 - 6 | 532 | 68 | ||
XM_018097082.1 | XP_017952571.1 | 5 - 6 | 576 | 68 | ||
XM_018097083.1 | XP_017952572.1 | 5 - 6 | 405 | 68 | ||
GenBank mRNA | BC075537.1 | - | 6 - 7 | 591 | 68 |
Gene Symbol: LOC101733964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR942529.1 | - | 5 - 6 | 428 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |