Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203651.1 | NP_988982.1 | 5 - 6 | 568 | 104 | |
GenBank mRNA | BC061339.1 | - | 5 - 6 | 568 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |