Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203688.1 | NP_989019.1 | 4 - 5 | 422 | 117 | |
GenBank mRNA | BC061434.1 | - | 4 - 5 | 422 | 117 | |
CR760193.2 | - | 4 - 5 | 381 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |