Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubqln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203890.1 | NP_989221.1 | 10 - 11 | 1813 | 64 | |
GenBank mRNA | BC063190.1 | - | 10 - 11 | 1813 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |