Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006908.1 | NP_001006909.1 | 4 - 5 | 335 | 103 | |
XM_012964546.2 | XP_012820000.1 | 4 - 5 | 346 | 103 | ||
GenBank mRNA | BC075325.1 | - | 4 - 5 | 335 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |