Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008100.1 | NP_001008101.1 | 3 - 4 | 412 | 76 | |
XM_012970334.2 | XP_012825788.1 | 4 - 5 | 547 | 76 | ||
XM_012970335.2 | XP_012825789.1 | 4 - 5 | 800 | 76 | ||
XM_018093859.1 | XP_017949348.1 | 3 - 4 | 505 | 76 | ||
GenBank mRNA | BC081302.1 | - | 3 - 4 | 412 | 76 | |
CR761852.2 | - | 3 - 4 | 324 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |