Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnni2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011110.2 | NP_001011110.1 | 6 - 7 | 343 | 92 | |
XM_012960980.1 | XP_012816434.1 | 6 - 7 | 390 | 92 | ||
XM_012960981.1 | XP_012816435.1 | 5 - 6 | 764 | 92 | ||
GenBank mRNA | BC084508.1 | - | 6 - 7 | 344 | 92 | |
CU464111.1 | - | 6 - 7 | 320 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |