Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015905.2 | NP_001015905.1 | 1 - 2 | 219 | 67 | |
GenBank mRNA | BC170741.1 | - | 1 - 2 | 200 | 67 | |
BC171252.1 | - | 1 - 2 | 200 | 67 | ||
CR926437.2 | - | 1 - 2 | 219 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |