Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030474.1 | NP_001025645.1 | 3 - 4 | 781 | 71 | |
XM_012957094.2 | XP_012812548.1 | 3 - 4 | 819 | 71 | ||
GenBank mRNA | BC091618.1 | - | 3 - 4 | 781 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |