Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030503.1 | NP_001025674.1 | 3 - 4 | 278 | 58 | |
GenBank mRNA | BC093459.1 | - | 3 - 4 | 278 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |