Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016299.2 | NP_001016299.1 | 2 - 3 | 449 | 66 | |
GenBank mRNA | BC170775.1 | - | 2 - 3 | 447 | 66 | |
BC170801.1 | - | 2 - 3 | 447 | 66 | ||
CR761526.2 | - | 2 - 3 | 449 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |