Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhbdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016449.2 | NP_001016449.1 | 5 - 6 | 798 | 79 | |
XM_012963006.2 | XP_012818460.1 | 5 - 6 | 866 | 79 | ||
GenBank mRNA | CR760998.2 | - | 5 - 6 | 798 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |