Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017023.2 | NP_001017023.1 | 3 - 4 | 335 | 117 | |
XM_018092101.1 | XP_017947590.1 | 3 - 4 | 479 | 117 | ||
XM_018092102.1 | XP_017947591.1 | 2 - 3 | 287 | 117 | ||
GenBank mRNA | BC167324.1 | - | 3 - 4 | 433 | 117 | |
CR762087.2 | - | 3 - 4 | 335 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |