Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203606.2 | NP_988937.1 | 6 - 7 | 453 | 98 | |
GenBank mRNA | BC061397.1 | - | 6 - 7 | 476 | 98 | |
CR761520.2 | - | 6 - 7 | 453 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |