Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078720.1 | NP_001072188.1 | 2 - 3 | 267 | 72 | |
XM_018090185.1 | XP_017945674.1 | 2 - 3 | 804 | 72 | ||
GenBank mRNA | BC118721.1 | - | 2 - 3 | 267 | 72 | |
CT025300.2 | - | 2 - 3 | 269 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() |