Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078793.1 | NP_001072261.1 | 2 - 3 | 1199 | 96 | |
XM_018092157.1 | XP_017947646.1 | 2 - 3 | 925 | 96 | ||
GenBank mRNA | BC121249.1 | - | 2 - 3 | 1199 | 96 | |
CT027965.3 | - | 2 - 3 | 1197 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |