Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078840.1 | NP_001072308.1 | 1 - 2 | 570 | 77 | |
GenBank mRNA | BC121365.1 | - | 1 - 2 | 570 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |