Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016858.4 | NP_001016858.3 | 3 - 4 | 911 | 60 | |
XM_012961143.2 | XP_012816597.1 | 3 - 4 | 1544 | 60 | ||
GenBank mRNA | BC135252.1 | - | 3 - 4 | 911 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |