Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fibcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079051.1 | NP_001072519.1 | 6 - 7 | 1490 | 74 | |
GenBank mRNA | BC121964.1 | - | 6 - 7 | 1490 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |