Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b3glct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079083.1 | NP_001072551.1 | 5 - 6 | 387 | 70 | |
XM_012957278.2 | XP_012812732.1 | 4 - 5 | 480 | 70 | ||
GenBank mRNA | BC122064.1 | - | 5 - 6 | 387 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |