Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102838.1 | NP_001096308.1 | 14 - 15 | 1465 | 71 | |
GenBank mRNA | BC135661.1 | - | 14 - 15 | 1465 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |