Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ric3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078773.1 | NP_001072241.1 | 2 - 3 | 336 | 86 | |
GenBank mRNA | BC118843.1 | - | 2 - 3 | 336 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |