Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twist2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103209.1 | NP_001096679.1 | 1 | 357 | 91 | |
XM_012956323.2 | XP_012811777.1 | 1 | 653 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |