Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112961.2 | NP_001106432.1 | 6 - 7 | 745 | 99 | |
XM_012969733.2 | XP_012825187.1 | 6 - 7 | 964 | 99 | ||
GenBank mRNA | EL802546.1 | - | 1 - 2 | 329 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |