Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gabpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123022.1 | NP_001116494.1 | 2 - 3 | 173 | 65 | |
GenBank mRNA | BC161465.1 | - | 2 - 3 | 173 | 65 | |
CT025182.2 | - | 2 - 3 | 132 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |